Learn More
Creative Biomart Recombinant H3N2 Antigen, His-tagged
Supplier:  Creative Biomart VAC0374
Encompass_Preferred
H3N2 produced in Hi-5 cell of Baculovirus is a single polypeptide chain containing 339 amino acids (17-345) and having a molecular mass of 37.8kDa.H3N2 is fused to a 6 amino acid His-tag at C-terminus
Catalog No. 50-208-5214
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.