Learn More
Creative Biomart Typhoid Vi polysaccharide vaccine Antigen
Supplier:  Creative Biomart VAC0357
Encompass_Preferred
Typhoid Vi polysaccharide vaccine Antigen is a vaccine that is administered intramuscularly. It is an active immunization for the prevention of typhoid fever caused by S typhi and is approved for use in perso
Catalog No. 50-208-5197
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.