Learn More
Creative Biomart Recombinant Influenza-A H1N1 New Caledonia Antigen
Supplier:  Creative Biomart VAC0227
Encompass_Preferred
Recombinant Full-Length H1N1 New Caledonia/20/99 is glycosylated with N-linked sugars, produced using baculovirus vectors in insect cells and its Mw is approximately 72 kDa.The insect cells infected w
Catalog No. 50-208-5067
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.