Learn More
Creative Biomart Recombinant HIV-1 Protease Antigen
Supplier:  Creative Biomart VAC0219
Encompass_Preferred
HIV-1 protease is an active homodimer having a molecular mass of 21.6 kDa (each monomer of 99 amino acids is 10.8 kDa).
Catalog No. 50-208-5059
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.