Learn More
Creative Biomart Recombinant HIV-1 TAT Antigen, Biotin-Labled
Supplier:  Creative Biomart VAC0220
Encompass_Preferred
HIV-1 TAT Biotin Recombinant-produced in E. coli is a single, non-glycosylated, polypeptide chain containing 86 amino acids encoded by two exons and having chain having a molecular mass of 14 kDa and
Catalog No. 50-208-5060
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.