Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A Virus H1N1 Hemagglutinin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB113961SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB113961SP 25 μg
RD10000243 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB113961SP Supplier R&D Systems Supplier No. MAB113961SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Hemagglutinin/HA Peptide Monoclonal antibody specifically detects Hemagglutinin/HA Peptide in Influenza A virus H1N1 samples. It is validated for Western Blot, Simple Western, Immunocytochemistry

Specifications

Antigen Hemagglutinin/HA Peptide
Applications Western Blot, Simple Western, Immunocytochemistry
Classification Monoclonal
Clone 2908F
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, Simple Western 25 μg/mL, Immunocytochemistry 3-15 μg/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
Gene Accession No. YP_009118626
Host Species Rabbit
Immunogen Human embryonic kidney cell, HEK293-derived influenza a virus H1N1 Hemagglutinin with a C-terminal 6-His tag, Met1-Gln529, Accession # YP_009118626
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Influenza A virus H1N1
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.