Learn More
Bioworld Influenza A Virus H1N1 Taiwan 1/86 (Recombinant), 10 μg
Supplier:  Bioworld 220605171
Encompass_Preferred
Influenza,type A virus (H1N1) Taiwan/1/86 Source: Allantoic fluid Description : Influenza A was harvested from Allantoic fluid of 10 days old embryonated eggs, inoculated with the influenza strain- Taiwan/1/86. Influenza type-A is purified by ultracentrifugation using 10-40% sucrose gradient. Formulation: Solution in STE containing 0.1% NaN3 and 0.005% merthiolate Inactivation: 0.005% merthiolate Storage: Influenza should be stored desiccated below -18 C. Please avoid freeze-thaw cycles. Purity: Greater than 90.0% as determined by: (c) Analysis by reducing and non-reducing SDS-PAGE Silver Stained gel. Dimers and aggregates: Less than 1% as determined by silver-stained SDS-PAGE gel analysis. Immunological activity: Has been tested with anti-influenza A monoclonal antibodies in ELISA. Usage: BioWORLD\'s products are furnished for LABORATORY RESEARCH USE ONLY. The product may not be used as drugs, agricultural or pesticidal products, food additives or household chemicals.
Catalog No. 50-253-615
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.