Learn More
ATCC Influenza A virus (H3N2) strain A/Baltimore/JH-335/2022 - ATCC VR-3398 (frozen viral culture)
Supplier:  ATCC VR3398
Encompass
Influenza A virus (H3N2) strain A/Baltimore/JH-335/2022 (ATCC VR-3398) is a characterized H3N2 influenza A isolate supplied as a frozen viral culture propagated in MDCK-SIAT1 cells. It is intended for research use in virology studies, vaccine research, antigenic characterization, and assay development.
  • Characterized H3N2 influenza A strain suitable for research applications.
  • Supplied as a frozen viral culture propagated in MDCK-SIAT1 cells.
  • Suitable for virology, vaccine research, and assay development.
  • Genomic sequence data available via ATCC Genome Portal.
  • Includes images showing host cell effects.
Catalog No. NC3585201