Learn More
ATCC Influenza A Virus (H3N2), Strain MRC 2 (ATCC VR-777)
Supplier: ATCC VR777
ATCC Influenza A virus (H3N2) VR-777 is a characterized viral isolate provided for research use. Strain designation MRC 2, this recombinant isolate derives from A/England/42/72 and A/PR8/34 lineages and has a published genome sequence. It is intended for virology research, antigenicity studies, and assay or diagnostic development when handled under appropriate biosafety conditions.
- Strain designation: MRC 2.
- H3N2 antigenic type with hemagglutinin and neuraminidase matching A/England/42/72.
- Genome sequenced and record available in public genome portal.
- Isolation source: human recombinant lineage.
- Recommended storage: -70°C or colder.
- Biosafety level 2; use appropriate personal protective equipment.
- Suitable for virology research and assay development.