Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Influenza B Hemagglutinin Antibody Pair [HRP]
SDP

Catalog No. NB397950 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
15 Plates
5 Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB397950 15 Plates
NB397949 5 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB397950 Supplier Novus Biologicals™ Supplier No. NBP31165715PLATES
Add to Cart
Add to Cart

Pair [HRP]

specifically detects in Virus samples. It is validated for ELISA∣Sandwich ELISA.

Specifications

Assay Range 62.5 to 4000 pg/mL (example only, lot dependent)
Assay Sensitivity 62.5 pg/mL (example only; lot dependent)
Conjugate HRP
Detection Method Sandwich ELISA
Immunoassay Kit Format Sandwich ELISA Kit
Product Type Sandwich ELISA
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Influenza B Hemagglutinin.
Target Species Virus
For Use With (Application) ELISA
Quantity 15 Plates
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym B1AG46
Target Virus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.