Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Influenza B NP Recombinant Rabbit Monoclonal Antibody (HL1073)
GREENER_CHOICE

Catalog No. PIMA542368
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIMA542368 100 μL
1 options
This item is not returnable. View return policy

Catalog No. PIMA542368

Supplier: Invitrogen™ MA542368

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial. For short-term storage (1-2 weeks), store at 4°C. For long-term storage, aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Influenza A and B are the two types of influenza viruses that cause epidemic human disease. Influenza B viruses currently circulating are divided into two antigenically and genetically distinct groups: the Victoria and Yamagata lineages. Influenza virus B is a genus in the virus family Orthomyxoviridae. A limited host range means that Influenza virus B pandemics are rare. The nucleoprotein protects the negative strand viral RNA from nucleases by encapsidating it. Encapsidated genomic RNA is termed the ribonucleoprotein (RNP) and serves as template for transcription and replication.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Influenza B NP
Applications ELISA, Lateral Flow, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL1073
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias Influenza B; Influenza NP
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of Influenza B virus Nucleoprotein (B/Taiwan/753/2005). The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.