Learn More
Creative Biomart Recombinant CoV-NL63 Antigen, His-tagged
Supplier:  Creative Biomart VAC0189
Encompass_Preferred
Recombinant Coronavirus NL63 c-terminal nucleoprotein produced in E. coli contains 221-340 amino acids and fused to a 6 a.a. His tag a C-terminal having a total of 130 a.a..
Catalog No. 50-208-5029
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.