Learn More
Creative Biomart Recombinant Chlamydia Pneumonia Antigen, His-tagged
Supplier:  Creative Biomart VAC0179
Encompass_Preferred
Recombinant Chlamydia pneumonia antigen, produced in E. coli is derived from VD2-VD3 region of the CP major outer membrane protein (this region is recognized only by Chlamydia pneumonia antibody), it
Catalog No. 50-208-5019
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.