Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Ingenol-3-angelate, PKC Activator, >98% Purity, Solid, 5 mg

Catalog No. 502705921
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5921 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5921 Supplier Abcam Supplier No. AB1442805MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Selective activator of PKC isoforms, such as PKCθ in T cells, with >98% purity.

  • Molecular Weight: 430.5 Da
  • Purity: >98%
  • Form: Solid
  • Application: Selective activator of PKC isoforms, such as PKCθ in T cells
  • Storage Buffer: 100% Ingenol-3-angelate
  • CAS Number: 75567-37-2
  • Storage Conditions: Store at -20°C

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Substrate Properties Chemical Substrate
Shipping Condition Ambient
Target Molecule Protein Kinase C (PKC)
Buffer 100% Ingenol-3-angelate
Content And Storage Store at -20°C.
Format Vial
Form Solid
Formulation 100% Ingenol-3-angelate
Quantity 5 mg
For Use With (Application) PKC activation
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.