Learn More
Ansh Labs Inhibin B Equin/Canin/Rat ELISA 96-wells, 15 L predilution Plasma/Serum, Range 6 calibrators ranging 5.2-758 pg/mL, LOD 2.3 pg/mL, Time 3.5 hours, Shelf Life 24 months
SDP
Supplier:  Ansh Labs AL163
Encompass_Preferred
Inhibin B (Equine, Canine, Mouse) ELISA has a total 3.5 hours incubation, calibrators and range of 6, 5.2-758 pg/mL, sensitivity of 2.3 pg/mL, sample of 15 µL predilution Serum. Reacts with Bovine, Canine, Equine, Mouse. Crossreactivity with related molecules is not significant.
Catalog No. 50-194-3740
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.