Learn More
STEMCELL Technologies GlyH-101, Size: 5 mg
SDP
Supplier:  STEMCELL Technologies 1000530
Encompass_Preferred
GlyH-101 is a cell-permeable glycine hydrazide that selectively and reversibly blocks the cystic fibrosis transmembrane conductance regulator (CFTR) channel (Ki = 4.3 µM; Sonawane et al.). The CFTR protein is a chloride anion channel involved in the secretion of fluid in many epithelial tissues, such as the airways and intestine (Ma et al.). Defects in the CFTR gene alter ion transport which can lead to cystic fibrosis (Dalli et al.; Ma et al.). CANCER RESEARCH· Blocks CFTR and inhibits cell division by inducing hyperpolarization in human gastric cancer cells (Zhu et al.).
Catalog No. 50-206-9342
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.