Learn More
Med Vet International Placadine (Medetomidine Hydrochloride) Sterile Injectable Solution 1mg/mL, 10mL, Each
Supplier:  Med Vet International PHPLACADINE10ML
Encompass_Preferred
Placadine Medetomidine Hydrochloride Sterile Injectable Solution is indicated for use as a sedative and analgesic in dogs over 12 weeks of age to facilitate clinical examinations, clinical procedures, minor surgical procedures not requiring muscle relaxation, and minor dental procedures where intubation is not required. The IV route of administration is more efficacious for dental care. See package insert for complete product details.
Catalog No. 50-282-8717
Due to product restrictions, please to purchase or view availability for this product.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.