Learn More
Thermo Scientific™ Injection Port Liners, Split/Splitless Mixed Liner Sample Pack
Shop All Thermo Scientific ProductsDescription
Optimize the performance of Agilent™ GC systems with our specifically designed Thermo Scientific™ Injection Port Liners for Agilent instruments. Highly deactivated and produced to exacting tolerances, these injection liners ensure exceptional reproducibility.
- Highly deactivated borosilicate glass
- Quartz wool packing material
- ID × OD × L: 4 × 6.3 × 78.5mm
Specifications
Specifications
| Product Type | Split/Splitless Mixed Liner Sample Pack |
| Quantity | 5 |
| For Use With (Application) | Split/Splitless |
| Description | 453AG001 |
| Length (Metric) | Mixed |
| Packing Material | Mixed |
| For Use With (Equipment) | Thermo Scientific TRACE 1300/1600 series and Agilent GC instruments |