Learn More
Restek™ Injection Port Weldments
SDP
Catalog No. 06611226 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
For Use With (Equipment):
Agilent 5890 GCs
Catalog No. For Use With (Equipment)
06-611-226 Agilent 5890 GCs
1 options
Catalog No. 06-611-226 Supplier Restek™ Supplier No. 22666
For Agilent GCs with Tekmar Purge and Trap Systems
  • Easily attach a purge-and-trap with pre-installed, low dead volume fittings
Specifications
Product Type GC
Injection Mode Split/Splitless
Description Split/Splitless Weldment
Requirements Use with Tekmar Purge and Traps
For Use With (Equipment) Agilent 5890 GCs