Learn More
Essendant INKCART220MLPRO4800LMG
Supplier:  Essendant EPST606C00
Encompass_Preferred
Epson UltraChrome K3 inks produce archival prints with amazing color fidelity and gloss level The inks are scratch resistant and provide consistently stable colors that significantly outperform lesser ink technologies Durable and easy to install This breakthrough ink technology also makes it the perfect choice for professional neutral and toned black and white prints with higher density levels and virtually no metamerism Device Types- Inkjet Printer Color(s)- Light Magenta Supply Type- Ink OEM/Compatible- OEM
Catalog No. 50-000-35604