Description
- Printing instruments use chemical-resistant ink and print directly on Superfrost/ColorFrost tab of the slide
- For histology, cytology, hematology, and microbiology applications
- Clipped 45° or straight 90° corners
- Frosted tabs in nine different colors
Specifications
Specifications
| Frosted | Tabs |
| Color | Lilac |
| Corner Style | 45° |