Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Inlet Liner, Straight Design (Unpacked)

Catalog No. 111004528 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
For Use With (Equipment):
PerkinElmer Auto System, Model 9000 Split/Splitless Capillary Injector
PerkinElmer Programmed Split/Splitless (PSS)
Quantity:
5 Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Quantity
11-100-4528 PerkinElmer Programmed Split/Splitless (PSS) 5 Pk.
11-101-0517 PerkinElmer Auto System, Model 9000 Split/Splitless Capillary Injector 5 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-100-4528 Supplier MilliporeSigma™ Supelco™ Supplier No. 2631405

The prime functions of the inlet liner are to limit sample degradation, improve vaporization, and to correctly position the syringe needle

  • Incorrect choice of an inlet liner causes higher potential of peak tailing and mass discrimination
  • Liner volume is recommended to be larger than the amount of sample injected by syringe so as to avoid any reaction with the sample
  • Suitable for making a connection between sample introduction and GC columns
  • Direct type has narrow bore which sustains a high linear velocity through the injection port
  • Diminishes band broadening and transferring all the samples to column, flash vaporization
TRUSTED_SUSTAINABILITY

Specifications

Packing Material None
Quantity 5 Pk.
Length (Metric) 86 mm
Product Type Inlet Liner
Deactivation Silanized
Description Straight Design
For Use With (Application) Trace Analytes
Diameter (Metric) Inner 1 mm
Diameter (Metric) Outer 4 mm
For Use With (Equipment) PerkinElmer Programmed Split/Splitless (PSS)
Liner Material Glass
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.