Learn More
Thermo Scientific INLET-OUTLET ASSY
Supplier:  Thermo Scientific 49089
Encompass_Preferred
Rotor; Centrifuge; Accessory; Inlet/Outlet Assembly for Continuous Flow Operation; Component of TZ-28 Continuous Flow Ultracentrifuge Conversion Kit (Thermo 12271); For use with TZ-28 Zonal Titanium Rotor
Catalog No. 50-867-107
Only null left
Add to Cart
Request a Quote