Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Inlet Tubes for Autoclavable Bottletop Dispenser

Catalog No. 1361517 Shop All Fisherbrand Products
Change view
Click to view available options
Length (Metric):
160 to 310 mm
175 to 340 mm
190 to 370 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
13-615-17 175 to 340 mm
13-615-16 160 to 310 mm
13-615-18 190 to 370 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 13-615-17
Add to Cart
Add to Cart
This item is not returnable. View return policy

For use with Fisherbrand Autoclavable Bottletop Dispenser. Inlet tube is used to transfer reagent from bottle to the dispenser. This tube is telescopic in nature, thus can be adapted for different sized bottles.

  • Made of FEP material
  • Telescopic tube; length is adjustable
  • Broader side: Outer dia. is 9mm, Inner dia. is 8mm
  • Narrower side: Outer dia. is 7mm, Inner dia. is 6mm
Note: Tube has a small end and a large end. The smaller end can be used with 2.5ml, 5ml and 10ml Dispensers. The larger end is good for 30ml, 60ml and 100ml Dispensers.

Certifications

CE Certified, ISO 13485-2003, ISO 9001-2008, Compliance as per ISO 8655

Specifications

Certifications/Compliance CE Certified, ISO 13485-2003, ISO 9001-2008, Compliance as per ISO 8655
Color Translucent
For Use With (Equipment) Fisherbrand Autoclavable Bottletop Dispenser
Length (Metric) 175 to 340 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.