Learn More
METTLER TOLEDO 3P INDUSTRIAL INMOTION KF PROOVEN AUTO 5ML
Supplier:  METTLER TOLEDO 3P INDUSTRIAL 30407503
Encompass
InMotion KF Pro Oven Autosampler (5mL)The InMotion KF autosampler is designed for water determination by the oven extraction technique.Upon heating to the sample-specific temperature, the water evaporates and is transferred into thetitration vessel by a stream of dry air or nitrogen.
Catalog No. NC2866700
Only null left
Add to Cart
Request a Quote