Learn More
Binder Inc Inner door divided CB 260
SDP
Supplier:  Binder Inc 86120031
Encompass_Preferred
Inner door, divided - Cell therapy compartmentalization, consisting of 8 inner doors and two shelf levels with four partition wall each - optional for BINDER CO2 incubator
Catalog No. 86-120-031
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.