Description
- All calibration certificates contain as-found and as-left data, estimated measurement uncertainties (emu's), and test uncertainty ratios (TUR's)
- NIST-traceable calibration allows users to confirm that the specific asset purchased meets the manufacturer's published specification
- EPA, USDA, ISO, and GLP/GMPs standards require regular NIST-Traceable calibration of all test equipment being used in critical applications, FDA compliant
- Product arrives calibrated and ready to use, eliminating the need to send it to a third-party or internal calibration lab and incurring additional delays
Specifications
Specifications
| For Use With (Equipment) | Flowmeter (Air/Gas non-Mass) |
| Certifications/Compliance | NIST-Traceable |
| Description | 0 to 500 LPM air flow capability |
| Calibration Points | 5 to 10 Measuring Points, (Meter Dependent) |
| Duration | 12 Months |
| Calibration Type | Flowmeter (Air/Gas non-Mass) |
| Product Type | Calibration |
Calibration reports issued by InnoCal may not be reproduced, except in full, without written permission of InnoCal. The results stated in our reports relate only to the items tested or calibrated.