Learn More
Innocal™ Humidity Calibrations
Catalog No. 1509431
Change view
Click to view available options
Product Type:
Humidity (High Accuracy)
Humidity (Psychrometer)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
15-094-31 Humidity (Psychrometer)
15-094-37 Humidity (High Accuracy)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 15-094-31 Supplier Innocal™ Supplier No. WD1700500
Only null left
Add to Cart
Add to Cart
Keeping humidity instruments calibrated with InnoCal™ Humidity Calibrations helps measure and regulate this variable more accurately.
  • All calibration certificates contain as-found and as-left data, estimated measurement uncertainties (emu's), and test uncertainty ratios (TUR's)
  • NIST-traceable calibration allows users to confirm that the specific asset purchased meets the manufacturer's published specification
  • EPA, USDA, ISO, and GLP/GMPs standards require regular NIST-Traceable calibration of all test equipment being used in critical applications, FDA compliant
  • Product arrives calibrated and ready to use avoiding the necessity of sending it to a third-party or internal calibration lab
  • All measurements reported are traceable to SI units via national standards maintained by NIST and performed in compliance with ANSI Z540, ISO17025, and MIL-STD-45662A.
Specifications
Certifications/Compliance NIST-Traceable
Description 0 to 95% RH capability
Duration 12 Months
Product Type Humidity (Psychrometer)
Includes Three test points across the range
Calibration reports issued by InnoCal may not be reproduced, except in full, without written permission of InnoCal. The results stated in our reports relate only to the items tested or calibrated.