Description
- NIST-traceable calibration allows users to confirm that the specific asset purchased does in fact meet the manufacturer's published specification
- EPA, USDA, ISO and GLP/GMPs standards require regular NIST-Traceable calibration of all test equipment being used in critical applications
- Product arrives calibrated and ready to use rather eliminating need to send to a third-party r internal calibration lab
- Calibration service for a bimetal or digital-pocket thermometer
Specifications
Specifications
| Certifications/Compliance | NIST-Traceable |
| Description | -196 to 1300°C capability |
| Duration | 12 Months |
| Calibration Type | Temperature (Bimetal or Digital Pocket) |
| Product Type | Calibration with Certificate and Data |
| Includes | ITS-90 points across range unless otherwise specified |
Calibration reports issued by InnoCal may not be reproduced, except in full, without written permission of InnoCal. The results stated in our reports relate only to the items tested or calibrated.