Learn More
Innovative Med Tech INNOSET Safety Blood Collection Sets with Luer Adapter
Catalog No. 22757170
Change view
Click to view available options
Needle Gauge:
21 G
25 G
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Gauge
22-757-170 21 G
22-757-172 25 G
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-757-170 Supplier Innovative Med Tech Supplier No. 596121
This item is not returnable. View return policy
Blood collection set with attached safety shield manually activated to cover the needle immediately after use to minimize risk of accidental needle stick by covering the used needle prior to disposal
Specifications
Product Type Blood collection Set with Male M/S Luer Adapter
Needle Gauge 21 G
Length (English) Needle 3/4 in.
Length (English) Tube 12 in.