Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eppendorf™ Innova™ 42R Shaker and Universal Platform Bundle PROMO
The Innova™ 42R refrigerated shaker plus 18 x 18 in. universal platform speeds plasmid prep, cell culture or enzyme screens. Stack units to free space and run multi-vessel workflows with ease.
Supplier: Eppendorf™ 2231001157
Description
Innova™ 42R shaker and Universal Platform Bundle Includes:
- 1 x Innova™ 42R with 3/4 in. or 1 in. orbit, 120V/60Hz
- 1 x Universal Platforms, 18 x 18 in.
Innova™ 42R Shaker Features:
- Height adjustable static shelf allows simultaneous shaking and static culture with different vessels
- Save space by stacking units or positioning them on or under a standard-height lab bench
- Benefit from configurable platforms that accommodate tubes, microplates, and flasks up to 6 L

Specifications
25 mm | |
25 to 400 rpm | |
120 V | |
12.5 A | |
Incubator Shaker | |
68°F to 176°F | |
60 Hz | |
29.3 in. | |
74.5 cm | |
51.5 x 51.7 x 47.7 cm (20.3 x 20.4 x 18.8 in.) | |
63.5 x 74.5 x 81.8 cm (25 x 29.3 x 32.8 in.) | |
18.8 in. | |
47.7 cm | |
20.4 in. | |
51.7 cm | |
121 kg | |
Innova 42R | |
20 to 80%, Non-condensing | |
0.01 sec. to 95 hr. 59 min., Continuous |
9 to 15.8 kg | |
120 V 50/60 Hz | |
20°C to 80°C | |
6 L Flasks | |
20 to 35 lb. | |
±0.25°C at 37°C | |
Aluminum | |
20.3 in. | |
51.5 cm | |
46 x 46 cm (18 x 18 in.) | |
32.2 in. | |
81.8 cm | |
25 in. | |
63.5 cm | |
266 lb. | |
±1 rpm | |
LED Digital Electric Display | |
Universal/Dedicated | |
Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction