Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ Innova™ 42R Shaker and Universal Platform Bundle PROMO

The Innova™ 42R refrigerated shaker plus 18 x 18 in. universal platform speeds plasmid prep, cell culture or enzyme screens. Stack units to free space and run multi-vessel workflows with ease.

Supplier:  Eppendorf™ 2231001156

 View more versions of this product

Catalog No. 05-400-157PM1


Get Up to 60% Off Eppendorf Equipment
View Available Promotions
Only null left
Add to Cart

Description

Description

Innova™ 42R shaker and Universal Platform Bundle Includes:

  • 1 x Innova™ 42R with 3/4 in. or 1 in. orbit, 120V/60Hz
  • 1 x Universal Platforms, 18 x 18 in.

Innova™ 42R Shaker Features:

  • Height adjustable static shelf allows simultaneous shaking and static culture with different vessels
  • Save space by stacking units or positioning them on or under a standard-height lab bench
  • Benefit from configurable platforms that accommodate tubes, microplates, and flasks up to 6 L
TRUSTED_SUSTAINABILITY
Specifications

Specifications

19 mm
25 to 400 rpm
120 V
12.5 A
Incubator Shaker
68°F to 176°F
60 Hz
29.3 in.
74.5 cm
51.5 x 51.7 x 47.7 cm (20.3 x 20.4 x 18.8 in.)
63.5 x 74.5 x 81.8 cm (25 x 29.3 x 32.8 in.)
18.8 in.
47.7 cm
20.4 in.
51.7 cm
121 kg
Innova 42R
20 to 80%, Non-condensing
0.01 sec. to 95 hr. 59 min., Continuous
9 to 15.8 kg
120 V 50/60 Hz
20°C to 80°C
6 L Flasks
20 to 35 lb.
±0.25°C at 37°C
Aluminum
20.3 in.
51.5 cm
46 x 46 cm (18 x 18 in.)
32.2 in.
81.8 cm
25 in.
63.5 cm
266 lb.
±1 rpm
LED Digital Electric Display
Universal/Dedicated
Yes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.