Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ InnovaCoat™ GOLD Antibody Labeling Kit, 80nm, Mini
SDP

Catalog No. 23100053RXN Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
3 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23100053RXN 3 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23100053RXN Supplier Novus Biologicals™ Supplier No. 23100053REACTIONS

Unique coated nanoparticle with a strongly bound protective surface coat

Specifications

Description InnovaCoat GOLD 80 nm Mini
Product Type Antibody Labeling Kit
Quantity 3 Reactions
Content And Storage Store at 4°C. Do not freeze.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.