Learn More
R&D Systems™ InnovaCoat (R) Gold - 10nm Midi Maleimide Antibody Labeling Kit
Shop All Novus Products

Description
This conjugation kit allows thiolated molecules (e.g. antibodies, Fab’ fragments, or other sulfhydryl-containing biomolecules) to be covalently attached to ultra-stable InnovaCoat™ GOLD nanoparticles at very high OD quickly and easily.
Kit Contents:
- 1 vial (1 reaction) of InnovaCoat™ GOLD Maleimide
- 1 vial of InnovaCoat™ GOLD Maleimide Reaction Buffer
- 1 vial of InnovaCoat™ GOLD Maleimide Quencher
- 1 vial or bottle of InnovaCoat™ GOLD Maleimide Diluent
Specifications
Specifications
| Target | InnovaCoat (R) Gold - 10nm Midi Maleimide |
| Content And Storage | -20°C |
| Product Line | Novus |
| Quantity | 400 μL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.