Learn More
R&D Systems™ InnovaCoat (R) GOLD - 40nm Midi Maleimide Antibody Labeling Kit
SDP
Catalog No. 2700015 Shop All Novus Products
Change view
Click to view available options
Quantity:
2.5 μg
Catalog No. Quantity
27-000-15 2.5 μg
1 options
For Research Use Only
Catalog No. 27-000-15 Supplier R&D Systems™ Supplier No. 2700015
Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.
For Research Use Only
A Novus product, part of R&D Systems. Use simple pipetting to covalently attach thiolated molecules to ultra-stable InnovaCoat™ GOLD nanoparticles. Simply add solution of thiolated biomolecule to the supplied freeze-dried mixture. Attached molecule isn'tt prone to dissociation, unlike many direct thiol-to-gold metal conjugations.
Features
  • Proprietary surface coat
  • Ultra-stable
  • Covalently link antibodies and analytes
  • Simple conjugation processes
  • Different surface chemistries
Benefits
  • Enhanced assay sensitivity
  • Protective coat prevents metal-antibody interactions
  • Rapid screening of multiple antibodies for assay development
  • Reduced antibody loading without aggregation
  • No pH titrations
  • Fully scalable – from R&D to production
  • Custom InnovaCoat™ labeling services
Specifications
Product Type Antibody Labeling Kit
Content And Storage -20°C
Product Line Novus
Quantity 2.5 μg