Learn More
Description
Order Info
Quantity
Specifications
Specifications
| Product Type | InnovaCoat GOLD 40nm Mini Antibody Labeling Kit |
| Content And Storage | 10 Vials of InnovaCoat GOLD, 1 Vial of Reaction Buffer. -20°C |
| Product Line | Novus |
| Quantity | 10 Tests |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
