Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ InnovaCoat (R) GOLD - 80nm Goat Anti-Rabbit IgG Gold (10OD)
SDP

Catalog No. 2210200 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2210200 0.2 mL
2211000 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 2210200 Supplier Novus Biologicals™ Supplier No. 2210200

Unique coated nanoparticle with a strongly bound protective surface coat

Specifications

Quantity 0.2 mL
Content And Storage Store at 4°C. Do not freeze.
Target InnovaCoat GOLD - 80nm Goat Anti-Rabbit IgG Gold (10OD)
Product Type Antibody Labeling Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.