Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ InnovaCoat (R) GOLD - 80nm Goat Anti-Rabbit IgG Gold (10OD)
SDP

Catalog No. 2211000 Shop All Novus Products
Change view
Click to view available options
Quantity:
0.2 mL
1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2211000 1 mL
2210200 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 2211000 Supplier R&D Systems™ Supplier No. 2211000

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Unique coated nanoparticle with a strongly bound protective surface coat

Specifications

Target InnovaCoat GOLD - 80nm Goat Anti-Rabbit IgG Gold (10OD)
Product Type Antibody Labeling Kit
Content And Storage Store at 4°C. Do not freeze.
Product Line Novus
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.