Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ InnovaCoat (R) GOLD 80nm Midi Antibody Labeling Kit
SDP

Catalog No. 2310015 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 Test
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-100-15 1 Test
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
For Research Use Only
Catalog No. 23-100-15 Supplier R&D Systems™ Supplier No. 2310015

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

For Research Use Only

A Novus product, part of R&D Systems. Use simple pipetting operations to covalently attach antibodies, proteins and other biomolecules to ultra-stable InnovaCoat GOLD nanoparticles. Hands-on time is about two minutes and the conjugate is ready within fifteen.

Features
  • Proprietary surface coat reduces risk of sub-optimal antibody performance
  • Ultra-stable
  • Covalently link antibodies and analytes
  • Simple conjugation processes
  • Different surface chemistries
Benefits
  • Antibody-GOLD conjugates in just 15 minutes
  • Enhanced assay sensitivity
  • Protective coat prevents metal-antibody interactions
  • Coating procedure not dependent on isoelectric point of antibody
  • Rapid screening of multiple antibodies for assay development
  • Reduced antibody loading without aggregation
  • No pH titrations
  • Fully scalable – from R&D to production
  • Custom InnovaCoat™ labeling services

Specifications

Product Type Antibody Labeling Kit
Content And Storage Store at 4°C. Do not freeze.
Product Line Novus
Quantity 1 Test
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.