Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ InnovaCoat (R) GOLD - Carboxyl 40OD 40nm
SDP

Catalog No. 34000001 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
200 μg
40 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-000-001 40 μg
34-000-005 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 34-000-001 Supplier Novus Biologicals™ Supplier No. 34000001

Coated gold nanoparticles functionalized with carboxyl groups that allow the covalent conjugation of antibodies, using the water soluble carbodiimide crosslinker EDC

Specifications

Quantity 40 μg
Content And Storage Store at 4°C. Do not freeze.
Target InnovaCoat GOLD - Carboxyl 40OD 40nm
Product Type Antibody Labeling Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.