Learn More
Siemens Healthineers INNOVANCE™ Antithrombin Kit
Catalog No. 23044736 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
100 Tests/Kit
130 Tests/Kit
450 Tests/Kit
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-044-736 450 Tests/Kit
23-044-737 100 Tests/Kit
23-313-003 130 Tests/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 23-044-736 Supplier Siemens Healthineers Supplier No. 10487303
Add to Cart
Add to Cart
This item is not returnable. View return policy
Chromogenic assay for the automated quantitation of functionally active antithrombin in human citrated plasma.
The INNOVANCE™ Antithrombin assay is an automated, ready-to-use, chromogenic assay that exhibits excellent precision and reliability. This quantitative assay for the determination of functional antithrombin utilizes human anti-Xa substrate and avoids interference with heparin cofactor II and direct thrombin inhibitors, such as hirudin.
TRUSTED_SUSTAINABILITY
Specifications
Type Hemostasis Kit
For Use With (Application) Activated Partial Thromboplastin Time (aPPT) Testing
Quantity 450 Tests/Kit