Learn More
Siemens Healthineers INNOVANCE™ Free Protein S Antigen Kit
Catalog No. 23044874 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
152 Tests/Kit
Catalog No. Quantity
23-044-874 152 Tests/Kit
1 options
This item is not returnable. View return policy
Catalog No. 23-044-874 Supplier Siemens Healthineers Supplier No. 10873458
Add to Cart
Add to Cart
This item is not returnable. View return policy
For the quantitative determination of free protein S antigen in human plasma collected from venous blood samples in 3.2% sodium citrate tubes on the BCS XP, Sysmex™ CS-2500 and Sysmex™ CS-5100 analyzers.
Free Protein S Antigen, a highly specific and stable assay for the quantitation of free Protein S antigen in human plasma.
TRUSTED_SUSTAINABILITY
Specifications
Type Hemostasis Kit
For Use With (Application) Thrombophilia Testing
Quantity 152 Tests/Kit
WARNING: Reproductive Harm - www.P65Warnings.ca.gov