Learn More
Siemens Healthineers INNOVANCE™ Heparin Assay (US)
Catalog No. 23044774 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
180 Tests/Kit
Catalog No. Quantity
23-044-774 180 Tests/Kit
1 options
This item is not returnable. View return policy
Catalog No. 23-044-774 Supplier Siemens Healthineers Supplier No. 10873535
Add to Cart
Add to Cart
This item is not returnable. View return policy
For the quantitative determination of the activity of unfractionated heparin (UFH) and low molecular weight heparin (LMWH).
INNOVANCE™ Heparin Assay quantitatively determines the activity of unfractionated (UF) and low-molecular-weight (LMW) heparin in citrated plasma. Liquid reagents and a single hybrid calibration curve for UF and LMW heparin help deliver precise results in fewer steps.
TRUSTED_SUSTAINABILITY
Specifications
Type Hemostasis Kit
For Use With (Application) Heparin Testing
Quantity 180 Tests/Kit