Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Blood Typing Anti-Sera
SCI_ED

Catalog No. S06625 Shop All Aldon Products
Change view
Click to view available options
Type:
Blood Typing Anti-Sera Type A, B and Rh
Blood Typing Anti-Sera: Anti Rh
Blood Typing Anti-Sera: Anti-A and Anti-B
Blood Typing Anti-Sera: Anti-H Lectin
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
S06625 Blood Typing Anti-Sera: Anti-A and Anti-B
S06626 Blood Typing Anti-Sera Type A, B and Rh
S06627 Blood Typing Anti-Sera: Anti Rh
S37733 Blood Typing Anti-Sera: Anti-H Lectin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S06625 Supplier Innovating Science™ Supplier No. IS3150
Add to Cart
Add to Cart

For blood typing using real human blood. 5 mL vials contain enough antisera for 75 students. Sample will keep over one year with proper refrigeration.

  • Five milliliter vial contains enough antisera for 75 students
  • Sample will keep over one year with proper refrigeration
  • Materials from human-derived cell lines were found to be negative in tests for Hepatitis B, HIV and EBV
  • Includes Anti-A & Anti-B, Anti Rh and Anti-H Lectin Anti-sera
  • DOT: Non regulated

Specifications

Content And Storage Ambient
CE Marker No
Concentration 1
Type Blood Typing Anti-Sera: Anti-A and Anti-B
For Use With (Application) For Blood typing of real Human Blood
Includes Anti-A and Anti-B and Anti Rh Anti-sera
Packaging Type Plastic Bottle
Quantity 1
Form Liquid

For educational use only. Not FDA approved for clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.