Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Cereal Grass Medium
GREENER_CHOICESCI_ED

Catalog No. S05868 Shop All Aldon Products
Change view
Click to view available options
Quantity: 25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S05868 Quantity: 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S05868 Supplier Innovating Science™ Supplier No. IS5020
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aldon’s Innovating Science™ cereal grass medium is used for culturing protozoa (Rhizopods, Choanoflagellates, Ciliates, and Flagellates). It contains dehydrated cereal grass leaves with natural vitamins A,B,C,K.

  • Contains dehydrated cereal grass leaves with natural vitamins A, B, C, and K
  • Recommended by King's lab at The University of California at Berkeley
  • DOT Info: Non-regulated

Specifications

Concentration 1
Content And Storage Ambient
Form Dehydrated
Manufacturing Origin United States
Media Type Cereal Grass Medium
Product Type Cereal Grass Medium
Sterility Non-sterile
Target Organism Class Protozoan Nucleic Acids
Test Type Growth Promotion Testing
For Use With (Application) Cultivation of protozoa (rhizopods, choanoflagellates, ciliates and flagellates)
Packaging Type Plastic Bottle
Quantity 25 g
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.