Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Freeze Dried Bacteria Cultures
SCI_ED

Catalog No. S07806 Shop All Aldon Products
Change view
Click to view available options
Product Type:
Bacillus subtilis
E. coli
Micrococcus luteus
Serratia marcescens
Staphylococcus epidermidis
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S07806 Bacillus subtilis
S07809 Serratia marcescens
S07807 E. coli
S07808 Micrococcus luteus
S07811 Staphylococcus epidermidis
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. S07806 Supplier Innovating Science™ Supplier No. IS3901
Add to Cart
Add to Cart

Includes

One tube of rehydrating broth and one tube agar slant, sterile swab and instructions.

The bacteria culture is freeze dried into a lyophilized pellet packaged in a sterile plastic vial. The culture includes one tube of rehydrating broth and one tube agar slant, sterile swab and instructions.

  • Lyophilized pellet form in vial
  • Available for five different types of bacteria

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) For use in classroom for educational purposes
Includes One tube of rehydrating broth and one tube agar slant, sterile swab and instructions
Quantity 1
Class Size 1 Class
Product Type Bacillus subtilis
Description Format: Lyophilized pellet
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.