Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Microbiology Science Fair Kit
GREENER_CHOICESCI_ED

Catalog No. s07805 Shop All Aldon Products
Change view
Click to view available options
Class Size:
15 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S07805 15 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S07805 Supplier Innovating Science™ Supplier No. IS5030
Add to Cart
Add to Cart

Enables students to grow their own bacteria by swabbing various surfaces in the classroom.

  • Hands-on, easy-to-use kit for students
  • Provides opportunity to investigate all phages of growing bacteria
  • Includes: 2g of nutrient agar (makes 100mL 2% Agar), six Petri dishes and six sterile swabs

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) For science fair, demonstrates to grow bacteria
Includes 2g nutrient agar (makes 100ml 2% agar), six petri dishes and six sterile swabs
Quantity 1
Dimensions (L x W x H) 5.5 x 4.25 x 6.75 in.
Class Size 15 Lab Groups
Product Type Microbiology Science Fair Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.