Learn More
Innovative Med Tech Blood Gas Sampling Syringes
Catalog No. p-3704258
Change view
Click to view available options
Quantity:
50 Pk.
Volume (Metric):
1 mL
Catalog No. Quantity Volume (Metric)
22-757-154 50 Pk. 1 mL
1 options
This item is not returnable. View return policy
Catalog No. 22-757-154 Supplier Innovative Med Tech Supplier No. 999017
Add to Cart
Add to Cart
This item is not returnable. View return policy
For laboratory or veterinary use only
  • Coated with approximately 50 I.U. balanced LI-heparin
  • Single use
  • Sterile
Specifications
For Use With (Application) Laboratory or veterinary use only
Product Type Blood Gas Sampling Syringe
Quantity 50 Pk.
Volume (Metric) 1 mL
Sterility Sterile