Learn More
Applied Biosystems™ InnoviGene™ Suite Software
Description
Applied Biosystems InnoviGene Suite software enables you to analyze and manage capillary electrophoresis sequencing and fragment analysis data generated by Applied Biosystems genetic analyzers. The software provides a streamlined workflow to guide you through uploading and analyzing the data and generating reports.
InnoviGene Suite includes SAE administrator console software with security, audit, and electronic signature (SAE) functions that support compliance with guidelines on electronic records security and prevents unauthorized access to the instrument.
The software is delivered via a download link sent to you after purchase.
Applications within InnoviGene Suite
InnoviGene Suite allows you to create projects for use with other applications that are integrated into the software. These applications enable you to complete an end-to-end workflow using projects that contain related files. The applications available depend on the type of InnoviGene Suite purchased ('sequencing' or 'fragment analysis').
Sequence Quality Check application (Innovigene Suite for sequencing)—enables you to perform the following activities:
- View the quality information for trace files
- Change quality settings and reanalyze trace files
- Perform comparative analysis across multiple trace files
Sequence Identity application (InnoviGene Suite for sequencing)—enables you to view the assembly of multiple files compared to a reference and view basecall mismatches. Specifically, you can perform the following activities with the application:
- Basecalling, quality value assignment, and mixed base identification
- Trimming low-quality ends from each sequence, if the setting is active
- Identifying poor-quality samples
- Assembling the samples against the reference sequence and generating a specimen consensus sequence
- Reviewing the basecalling quality values and consensus quality values
- Identifying mismatches by aligning and comparing specimen sequences to the reference sequence. A mismatch is a position in the specimen that does not match with the reference sequence. Mismatches include base changes, insertions, and deletions.
- Assigning specimen status based on alignment with the reference sequence and coverage criteria
- Editing basecalls in samples or the consensus
- Generating reports and results for export in CSV format
STR Typing application (InnoviGene Suite for fragment analysis)—the STR Typing application is genotyping software that uses short tandem repeat (STR) data. It enables you to perform the following activities:
- Analyze a mixture of DNA fragments, separated by size
- Provide a profile of the separation
- Calculate the sizes of the fragments
- Determine the alleles present in fragment analysis sample (FSA) files
Specifications
Specifications
| Sequencing Type | Sanger Sequencing |
| For Use With (Application) | Sanger Sequencing |
| Quantity | 5 Users |
| Product Line | InnoviGene |
| Format | Software |
| For Use With (Equipment) | Applied Biosystems™ Genetic Analyzers |
| License | 3 Years |
| Product Type | Sequencing Software |
Frequently Asked Questions (FAQs)
SeqScape templates and RDGs cannot be imported into the InnoviGene Suite. If you have a GenBank file (.gb file) with non-overlapping ROIs, it can be imported into the InnoviGene Suite. Please note that some features in .gb files may be ignored by the InnoviGene Suite. For detailed information on .gb file compatibility, please refer to the relevant sections in the Getting Started Guide (GSG) or User Guide (UG) that can be found here: https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN1000299-InnoviGeneSuiteSoftware-EFI.pdf and Capillary Electrophoresis Software Support Center.
The InnoviGene Suite data is stored on your local computer in a folder location specified by the user during the installation of InnoviGene Suite software. If another member of your group is added to the same SAE Administrator Console, the InnoviGene Suite allows you to share items with those same users within the InnvoiGene Suite. If they are using a different SAE Administrator Console License, trace files, projects, and other files can be exported and later imported into another InnoviGene Suite instance and Capillary Electrophoresis Software Support Center.
.The InnoviGene Suite does not have direct access to ThermoFisher Connect. With browser-based access, only colleagues on the same network and with user credentials on the same instance of the InnoviGene Suite may have access to the data. During installation of InnoviGene Suite software, the user will specify a folder location where all project data will be stored. Users will have to export files from the InnoviGene Suite and send to Thermo Fisher Scientific personnel, for troubleshooting purposes or data review. and Capillary Electrophoresis Software Support Center.
Co-installation with InnoviGene Suite software was tested for the following software:
- GeneMapper v6.1
- Sequencing Analysis v8.0 (without the SDB license)
- SeqScape v4.1
- Variant Reporter v3.1
- Plate Manager v2.1.0
Note: There is a known FlexNet licensing conflict between InnoviGene Suite and the SeqA 8 + SDB license and these softwares should not be co-installed. and Capillary Electrophoresis Software Support Center.
The InnoviGene Suite is compatible with the following various antivirus software:
- Symantec™ Endpoint Protection
- McAfee®Endpoint Security
- Norton Internet Security™
- Microsoft® Defender antivirus software and Capillary Electrophoresis Software Support Center.
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.