Learn More
Leica Biosystems Inoculating Loop 24 Ga., 3mm
Supplier:  Leica Biosystems 39101039
Encompass_Preferred
Inoculating Loops are calibrated for culturing, transferring, streaking or isolating tissue cultures in cytology or histology. The loop allows an inoculum to be taken from a liquid by using surface tension. The Inoculating portfolio comes in an array of diameters, gauges, and lengths.
Catalog No. 50-255-2400
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.